Learn More
Thermo Scientific™ VersaTREK™ Connectors
Catalog No. y715044
Click to view available options
Packaging Type:
50/Box
This item is not returnable. View return policy
Catalog No. Y715044
Supplier: Thermo Scientific™ Y715044
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Facilitate organism detection by linking culture bottle pressure changes to the Thermo Scientific™ VersaTREK™ instrument sensor while preventing gas build up in the bottles.
Facilitate organism detection by linking culture bottle pressure changes to the Thermo Scientific™ VersaTREK™ instrument sensor using Thermo Scientific VersaTREK Connectors. The recessed probe also prevents dangerous gas build up within the Thermo Scientific™ VersaTREK™ REDOX™ or Thermo Scientific VersaTREK Myco bottle.
Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.
  • Only system capable of detecting any gas produced or consumed by organisms, allowing detection of a wider range of both common and fastidious organisms
  • Simplified workflow includes easy scan function for bottle entry, intuitive icons to reduce training time, and no daily QC requirement for maximum efficiency
  • Highly-enriched Thermo Scientific VersaTREK REDOX media supports a wide range of organisms, facilitating better recovery of fastidious organisms
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description VersaTREK Connectors
Product Type Microbiology Media
Test Type Microbial Detection
No. per Pack 50
Packaging Type 50/Box
Specimen Type Various
Species M. avium ssp. paratuberculosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.