Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VersaTREK™ REDOX™ EZ Draw™ Media

Catalog No. p-6111367
Change view
Click to view available options
Product Type:
Microbiology Media
Media Type:
Aerobic media
Anaerobic media
Quantity:
40 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Media Type Quantity
Y710644 Microbiology Media Aerobic media 40 mL
Y710744 Microbiology Media Anaerobic media 40 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. Y710644 Supplier Thermo Scientific™ Supplier No. Y710644
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform safe, simple and convenient phlebotomy, without the need for costly blood adapters, using the only FDA-cleared blood culture bottles for true direct draws.

Perform safe, simple and convenient phlebotomy, without the need for costly blood adapters, with Thermo Scientific™ VersaTREK™ REDOX™ EZ Draw™ Media, the only FDA-cleared true, direct measured draw in the industry. Accommodating up to 5mL of blood, EZ Draw media is available in a 40mL broth for both REDOX 1 aerobic and REDOX 2 anaerobic bottles and can be used for manual or automated monitoring of direct-draw blood samples. Plus, the highly-enriched REDOX media supports a wide range of organisms, enhancing the fully automated VersaTREK System's ability to better recover fastidious organisms.

Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.
  • Easy and convenient sampling
  • Reduced risk from sharps by eliminating the need for syringe, butterfly, angel wings and other blood transfer devices
  • Reduced costs by eliminating the need for separate collection devices
  • Largest blood-to-broth dilution ratio in the industry (1:9) to dilute out the effects of antimicrobials, serum host factors and other agents
  • Enhanced, enriched media to support the wide range of organisms found in the microbiology laboratory
  • Available in 40 and 80mL sizes and suitable for all patient populations
Not all products are available for sale in all territories. Please inquire.

VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store away from direct sunlight
Media Type Aerobic media
Product Type Microbiology Media
Target Organism Class Aerobic Microorganisms
Test Type Microbial Detection
Manual/Automated Manual and Automated
No. per Pack 50 Bottles/Box
Quantity 40 mL
Specimen Type Blood, Sterile Body Fluid
Volume (Metric) Specimen 0.1 to 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.