Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ VeriShield™ 100 Series Over-the-Head Dielectric Ear Muffs

Catalog No. 19085528 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
CSA Z94.2:
Class A
Class AL
Foldable:
No
Yes
Noise Reduction Rating (NRR):
23 dB
25 dB
26 dB
29 dB
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2 Foldable Noise Reduction Rating (NRR)
19-085-528 Class A No 26 dB
19-085-525 Class A No 23 dB
19-085-527 Class A Yes 25 dB
19-085-530 Class AL No 29 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-085-528 Supplier Honeywell™ Supplier No. 1035192VS
Add to Cart
Add to Cart
This item is not returnable. View return policy

VeriShield™ Over-the-Head Dielectric Ear Muffs feature a durable, non-deforming plastic headband and are ideal for electrical environments.

  • Dielectric suitable for electrical environments
  • Durable plastic headband
  • Steel-wire construction with lightweight cushioned headband and precision micro adjustments are designed for excellent fit around the ear
  • Memory foam ear cushions combine dense and soft foam
  • Reduces squeezing pressure on the head and enhances comfort
  • Offered in low level, mid level or high level models

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
High Visibility No
Model VS120D
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Product Type VeriShield
Headband Color Black
Dielectric Yes
Style Over-the-head
Foldable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.