Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Versican, Alpha Domain Rabbit anti-Rat, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABT135825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT1358-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT1358-25UL Supplier MilliporeSigma Supplier No. ABT135825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Versican, Alpha Domain, Cat. No. ABT1358, is a rabbit polyclonal antibody that detects versican core protein that has been tested for use in Immunohistochemistry (Paraffin), Radioimmunoassay, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Versican, Alpha Domain
Applications Immunohistochemistry (Paraffin), Immunoradiometric Assay, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Chondroitin sulfate proteoglycan core protein 2;Glial hyaluronate-binding protein;GHAP;Large fibroblast proteoglycan;PG-M
Gene Symbols Vcan;Cspg2
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to the first 321 amino acids of rat Versican.
Molecular Weight of Antigen 300.01 kDa calculated.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects versican in rat eye sections.
Target Species Rat
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.