Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Verso cDNA Synthesis Kit

Catalog No. AB1453A
Change view
Click to view available options
No. of Reactions:
100 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
AB1453A 40 Reactions
AB1453B 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB1453A Supplier Thermo Scientific™ Supplier No. AB1453A
Add to Cart
Edge
Add to Cart

The Thermo Scientific Verso cDNA Synthesis Kit provides robust transcription of RNA to create a complete cDNA pool.

The Thermo Scientific Verso cDNA Synthesis Kit provides robust transcription of RNA to create a complete cDNA pool. The Verso system achieves robust and sensitive reverse transcription through the combination of a high affinity RT enzyme, a unique RNA priming method, and an optimized buffering system. The Verso RT enzyme has high RNA template affinity and reduced RNase H activity, to transcribe even sections with high secondary structure. The included anchored oligo dT priming method further enhances sensitivity by increasing transcription efficiency, and the cDNA synthesis buffer has been optimized to achieve a full and diverse cDNA pool.

Features of the Verso cDNA Synthesis Kit include:
• Highly sensitive with a broad dynamic range—can reverse transcribe RNA from an input range of 1pg to 1μg
• Efficient RT reaction allows for 75% shorter protocol times (down to 30 minutes)
• Unique priming strategy for increased efficiency and broad template flexibility
• High affinity RT enzyme effectively transcribes through difficult RNA sequences
• Wide working temperature range (42–57°C) for success with GC-rich and other difficult templates
• Included RT enhancer prevents genomic DNA carryover, eliminating the need for separate DNase I treatment
• RNase inhibitor included
• Can be paired with any PCR Kit for complete 2-Step RT-(q)PCR

Applications
• 2-step RT-(q)PCR
• Single stranded cDNA synthesis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• RT Enzyme mix with RNase Inhibitor
• 5X RT Buffer
• Random Hexamers
• Anchored Oligo (dT)
• 20 mM dNTPs
• RT Enhancer

Store at –20°C.

Format Kit
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 40 reactions
Reverse Transcriptase Verso
Ribonuclease H Activity Yes
Shipping Condition Wet or Dry Ice
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 40 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 11 kb
Starting Material RNA
Show More Show Less
What is the RT Enhancer that is included in all Verso kits?

The RT Enhancer is a unique enzyme that degrades contaminating genomic DNA at the start of the RT step. It is equally as effective as DNase I treatment but more convenient as it eliminates the need for a separate DNase I incubation prior to RT-PCR.

Do I need to treat my RNA with DNase I before running RT-qPCR with Verso cDNA Synthesis Kit?

No, you do not need to treat your RNA with DNase I before running RT-qPCR with the Verso cDNA Synthesis Kit. The RT enhancer included in the Verso kit degrades any contaminating double-stranded DNA during the reverse transcription process, eliminating the need for a separate DNase I treatment.

What is the size of cDNA that can be obtained from the Verso cDNA Synthesis Kit (Cat. No. AB1453B)

The Verso can synthesize long cDNA strands, up to 11 kb, at a temperature range of 42 degrees C to 57 degrees C. For improved performance, we recommend SuperScript IV Reverse Transcriptase.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.