Learn More
Cytiva Vertical Gel Unit Spacer
Catalog No. 45002439 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Vertical Gel Unit
Catalog No. For Use With (Equipment)
45-002-439 Vertical Gel Unit
1 options
Catalog No. 45-002-439 Supplier Cytiva Supplier No. 80618089
Add to Cart
Add to Cart
Accessory for SE 600 Ruby Vertical Electrophoresis System.
  • For vertical gel unit spacer
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vertical Gel Unit
Type Spacer
Quantity 2/Pk.
Thickness (Metric) 1.5 mm
Width (Metric) 2 cm
Height (Metric) 16 cm