Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Verubecestat
GREENER_CHOICESDP

Catalog No. 765950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-595-0 50 mg
76-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-595-0 Supplier Tocris Bioscience™ Supplier No. 7659/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BACE1 and BACE2 inhibitor

Verubecestat is a high affinity and selective β-secretase 1 (BACE1) and its homolog BACE2 inhibitor (Ki of 2.2 nM and 0.38 nM respectively). It exhibits no inhibition of cathepsin D, cathepsin E, pepsin, or renin. Verubecestat potently reduces Aβ40 formation in HEK293 APPSwe/Lon cells (IC50 = 2.1 nM). Verubecestat significantly lowers CSF and brain Aβ levels in rats. Orally bioavailable.

Specifications

Chemical Name or Material N-[3-[(5 R)-3-Amino-5,6-dihydro-2,5-dimethyl-1,1-dioxido-2 H-1,2,4-thiadiazin-5-yl]-4-fluorophenyl]-5-fluoro-2-pyridinecarboxamide
CAS 1286770-55-5
Quantity 50 mg
Target Secretase Inhibitors
Molecular Formula C17H17F2N5O3S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.