Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Vesicular Glutamate Transporter 3 (VGLUT3), Guinea Pig, Unlabeled, Polyclonal,

Catalog No. AB5421IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB5421IMI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5421IMI Supplier MilliporeSigma™ Supplier No. AB5421I
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Low-density lipoprotein receptor (UniProt P01130; also known as LDL receptor) is encoded by the LDLR (also known as FH, FHC) gene (Gene ID 3949) in human. Low-density lipoprotein (LDL) receptor recognizes the apoprotein B100 embedded in the outer phospholipid layer of cholesterol-rich LDL particles and mediates their remval by endocytosis via clathrin-coated pits. LDL receptor also recognizes the apoE protein found in chylomicron remnants and VLDL remnants (IDL). LDLR gene mutations are known causes of familial hypercholesterolaemia (FH). LDL receptor is initially produced with a signal peptide sequence (a.a. 1-21), the removal of which yields the mature single transmembrane (a.a. 789-810) protein wiht a C-terminal cytoplasmic tail (a.a. 811-860) and a large N-terminal extracellular region (a.a. 22-788) that contains seven class A (a.a. 25-313) and six class B (a.a. 397-658) LDLR repeats, as well as three EGF-like domains (a.a. 314-353, 354-393, 663-712). LDL-A repeats mediate LDL binding, repeats 2-7 are required for ApoB binding, while only repeat 5 is required for ApoE binding. The class B (LDL-B) repeats region adopts a beta-propeller conformation and is believed to be responsible for the pH-dependent release of LDL in the endosome. The cytoplasmic tail contains a signal sequence important for localizing the receptors to clathrin-coated pits and for triggering receptor-mediated endocytosis upon LDL binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vesicular Glutamate Transporter 3 (VGLUT3)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Guinea pig polyclonal antibody serum with 0.05% Sodium Azide.
Gene Slc17a8, Vglut3
Gene Accession No. Q7TSF2
Host Species Guinea Pig
Immunogen KLH-conjugated linear peptide corresponding to a C-terminal cytoplasmic domain sequence of rat vesicular glutamate transporter 3 (VGluT3).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_714947
Test Specificity This polyclonal guinea pig antiserum was raised against a VGluT3 C-terminal cytoplasmic domain sequence conserved between rat and murine species and shares 96% homology with the corresponding human sequence.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.