Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS Vesphene™ III st Disinfectant
Shop All STERIS Corporation Products
Click to view available options
Container Type:
Bottle
Description
- A liquid concentrate specifically formulated for the disinfection of hard surfaces
- Kills a broad spectrum of common bacteria and fungi at a use-dilution of rate of 0.8% v/v (1:128)
- Phenolic active ingredients provide broad spectrum antimicrobial efficacy
- Formulation can be used alone, or in rotation with the LpH™ III Disinfectants
- Gamma irradiated and double bagged for easy introduction into aseptic areas
- Products include an extensive documentation package to meet validation objectives
- Supported by the STERIS Technical Services team for individualized application and validation assistance.
- Each lot documented by Certificate of Analysis
Specifications
Specifications
| Product Type | Phenolic Disinfectant |
| Form | Concentrated Liquid |
| Sterility | Sterile |
| Concentration | 0.8% v/v (1:128) |
| For Use With (Application) | Disinfection of Hard Surfaces |
| Quantity | 144/Cs. |
| Volume (English) | 2 oz. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction