Learn More
Stoelting™ Vessel Cannulation Forceps Dumont INOX
SDP
Stoelting™ Vessel Cannulation Forceps are useful for introducing a small plastic tube into a blood vessel of almost equal size.
$598.00 - $715.50
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
10-001-145
SDP
View Documents
Stoelting™
5210065
0.35 mm OD
Each for $715.50
Only null left
Add to Cart
10-001-146
SDP
View Documents
Stoelting™
5210066
0.05 mm OD
Each for $715.50
Only null left
Add to Cart
10-001-147
SDP
View Documents
Stoelting™
5210067
1 mm OD
Each for $598.00
Only null left
Add to Cart
Description
Description
Hollowed jaws hold tubing securely without deforming it and the tip is guided into the vessel opening while preventing damage from unwanted movement.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.