Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay

Catalog No. p-7180675
Encompass_Preferred
Click to view available options
No. of Reactions:
100
500
This item is not returnable. View return policy
This item is not returnable. View return policy

The VetMAX™ Xeno™ Internal Positive Control (IPC) - LIZ™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results.

The Xeno IPC LIZ assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.

Features of the VetMAX Xeno IPC - LIZ Assay include:

  • Provides confidence that qPCR test results are accurate and actionable
  • Easily integrates into any workflow
  • Greatly reduces the likelihood of false negatives

The VetMAX Xeno IPC - LIZ Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno™ Internal Positive Control RNA or VetMAX™ Xeno™ Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay (Cat. No. A29765) is also available.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 110 μL Xeno LIZ Primer Probe Mix. Store at -5°C to -30°C.
Format Kit
For Use With (Application) Detection and Identification of Animal Disease
Includes Internal Positive Control Assay
Label or Dye LIZ
No. of Reactions 100
Product Line VetMAX™, Xeno™
Product Type Internal Positive Control
Quantity 100 reactions
Shipping Condition Dry Ice
Species Not Species-Specific
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.