Learn More
Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control RNA
Description
Xeno IPC RNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health PCR workflow.
Features of VetMAX Xeno IPC RNA include:
- Provides confidence that PCR test results are accurate and actionable
- Easily integrates into any workflow
- Greatly reduces the likelihood of false negatives
VetMAX Xeno IPC RNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.
Xeno IPC RNA is a component of our licensed VetMAX Gold detection kits, is recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC DNA is also available (Cat. No. A29762).
Specifications
Specifications
| Content And Storage |
|
| Format | Kit |
| For Use With (Application) | Detection and Identification of Animal Disease |
| Includes | Internal Positive Control |
| No. of Reactions | 500 |
| Product Line | VetMAX™, Xeno™ |
| Product Type | Internal Positive Control RNA |
| Quantity | 500 reactions |
| Shipping Condition | Dry Ice |
| Species | Not Species-Specific |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.