Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay

Catalog No. p-7180676
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A29767 500 Reactions
A29765 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A29767 Supplier Applied Biosystems™ Supplier No. A29767
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The VetMAX™ Xeno™ Internal Positive Control (IPC) - VIC™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results.

The Xeno IPC assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.

Features of the VetMAX Xeno IPC - VIC Assay include:

  • Provides confidence that qPCR test results are accurate and actionable
  • Easily integrates into any workflow
  • Greatly reduces the likelihood of false negatives

The VetMAX Xeno IPC - VIC Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno Internal Positive Control RNA or VetMAX™ Xeno Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay (Cat. No. A29766) is also available.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 550 μL Xeno VIC Primer Probe Mix. Store at -5°C to -30°C.
Format Kit
Label or Dye VIC
Product Line VetMAX, Xeno
Product Type Internal Positive Control
Quantity 500 reactions
Shipping Condition Dry Ice
Species Not Species-Specific
For Use With (Application) Animal Health
No. of Reactions 500 Reactions
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.