Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vezatin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572819
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572819 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572819 Supplier Thermo Scientific Supplier No. PA572819
Add to Cart
Add to Cart

Vezatin Polyclonal Antibody

Vezatin Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vezatin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene VEZT
Gene Accession No. Q3ZK22, Q5XI52, Q9HBM0
Gene Alias A630071D13Rik; AI848282; transmembrane protein vezatin; vezatin; vezatin, adherens junctions transmembrane protein; VEZT
Gene Symbols VEZT
Host Species Rabbit
Immunogen VEZT antibody for a 17 amino acid peptide near the center of human VEZT. The immunogen is within amino acids 480 - 530 of VEZT.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215008, 299738, 55591
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.