Learn More
Invitrogen™ VEZF1 Polyclonal Antibody

Description
Immunogen sequence: RLWEEAVKAR KKEAANLCQT STAATTPVTL TTPFSITSSV SSGTMSNPVT VAAAMSMRSP VNVSSAVNIT SPMNIGHPVT ITSPLSMTSP LTLTTPVNLP TPVTAPVNIA HPVTITSPMN LPTPMTLAAP LNIAMRPVES Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
Specifications
| Antigen | VEZF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | VEZF1 |
| Gene Accession No. | Q14119 |
| Gene Alias | AI848691; AW046909; DB1; putative transcription factor DB1; Vascular endothelial zinc finger 1; VEZF1; Zinc finger protein 161; ZNF161 |
| Gene Symbols | VEZF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.