Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VEZF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103478 Supplier Invitrogen™ Supplier No. PA5103478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total VEZF1.

VEZF1 is a possible transcription factor. It specifically binds to the CT/GC-rich region of the interleukin-3 promoter and mediates tax transactivation of IL-3.Transcriptional regulatory proteins containing tandemly repeated zinc finger domains are thought to be involved in both normal and abnormal cellular proliferation and differentiation. ZNF161 is a C2H2-type zinc finger protein. See MIM 603971 for general information on zinc finger proteins.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEZF1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VEZF1
Gene Accession No. Q14119
Gene Alias AI848691; AW046909; DB1; putative transcription factor DB1; Vascular endothelial zinc finger 1; VEZF1; Zinc finger protein 161; ZNF161
Gene Symbols VEZF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human VEZF1(Accession Q14119), corresponding to amino acid residues N290-Q340.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22344, 287615, 7716
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.