Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VG5Q Polyclonal Antibody
GREENER_CHOICE

Catalog No. APSA6599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APSA6599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APSA6599 Supplier Invitrogen™ Supplier No. PA316599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VG5Q Polyclonal Antibody for Western Blot, IHC (P)

VG5Q (Aggf1) gene encodes an angiogenic factor that promotes proliferation of endothelial cells. Mutations in this gene are associated with a susceptibility to Klippel-Trenaunay syndrome. Pseudogenes of this gene are found on chromosomes 3, 4, 10 and 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VG5Q
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene AGGF1
Gene Accession No. Q8N302
Gene Alias 2010009L17Rik; 2310029P06Rik; AGGF1; Angiogenic factor VG5Q; angiogenic factor with G patch and FHA domains 1; angiogenic factor with G-patch and FHA domains 1; AW112072; FLJ10283; G patch domain-containing protein 7; GPATC7; GPATCH7; HSU84971; HUS84971; hVG5Q; mVG5Q; Peg3; RGD1310888; Vasculogenesis gene on 5q protein; Vg5q
Gene Symbols AGGF1
Host Species Rabbit
Immunogen A synthetic peptide within residues 500-600 of the human VG5Q protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55109
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.