Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VG5Q Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987790
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7790 Supplier Bioss Supplier No. BS7509R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VG5Q (Aggf1) gene encodes an angiogenic factor that promotes proliferation of endothelial cells. Mutations in this gene are associated with a susceptibility to Klippel-Trenaunay syndrome. Pseudogenes of this gene are found on chromosomes 3, 4, 10 and 16.

Specifications

Antigen VG5Q
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AGGF1
Gene Accession No. Q8N302
Gene Alias 2010009L17Rik; 2310029P06Rik; AGGF1; Angiogenic factor VG5Q; Angiogenic factor with G patch and FHA domains 1; angiogenic factor with G-patch and FHA domains 1; AW112072; FLJ10283; G patch domain-containing protein 7; GPATC7; GPATCH7; HSU84971; HUS84971; hVG5Q; mVG5Q; Peg3; RGD1310888; Vasculogenesis gene on 5q protein; Vg5q
Gene Symbols AGGF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VG5Q/AGGF1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55109
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.