Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VG5Q Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | VG5Q |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
VG5Q Polyclonal specifically detects VG5Q in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
VG5Q | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Angiogenic factor VG5Q, angiogenic factor with G patch and FHA domains 1, FLJ10283hVG5Q, GPATC7G patch domain-containing protein 7, GPATCH7HUS84971, HSU84971, VG5QVasculogenesis gene on 5q protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human VG5Q (NP_060516.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Angiogenesis, Hypoxia | |
PBS with 50% glycerol, pH7.3. | |
55109 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title