Learn More
Invitrogen™ VGAT Polyclonal Antibody

Description
Positive Samples: Mouse spinal cord, Mouse brain; Cellular Location: Cytoplasmic vesicle membrane, Multi-pass membrane protein Immunogen sequence: MATLLRSKLS NVATSVSNKS QAKMSGMFAR MGFQAATDEE AVGFAHCDDL DFEHRQGLQM DILKAEGEPC GDEGAEAPVE GDIHYQRGSG APLPPSGSKD QVGGGGEFGG HDKPKITAWE AGWNVTNAIQ GMFVLGLPYA ILHGGYLGLF LIIFAAVVCC YTGKILIACL YEENEDGEVV RVRDSYVAIA NACCAPRFPT LGGRVVNVAQ IIELVMTCIL YVVVSGNLMY NSFPGLPVSQ KSWSIIATAV LLPCAFLKNL KAVSKFSLLC TLAHFVINIL VIAYCLSRAR DWAWEKVKFY IDVKKFPISI GIIVFSYTSQ IFLPSLEGNM QQPSEFHCMM NWTHIAACVL KGLFALVAYL TWADETKEVI TDNLPGSIRA VVNIFLVAKA LLSYPLPFFA AVEVLEKSLF QEGSRAFFPA CYSGDGRLKS WGLTLRCALV VFTLLMAIYV PHFALLMGLT GSLTGAGLCF LLPSLFHLRL LWRKLLWHQV FFDVAIFVIG GICSVSGFVH SLEGLIEAYR TNAED.
Specifications
Specifications
| Antigen | VGAT |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.79 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC32A1 |
| Gene Accession No. | O35458, O35633, Q9H598 |
| Gene Alias | GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; R75019; rGVAT; SLC32A1; solute carrier family 32 (GABA vesicular transporter), member 1; solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat |
| Gene Symbols | SLC32A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.