Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGCNL1 Antibody (S187-7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222410F
Description
VGCNL1 Monoclonal specifically detects VGCNL1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
VGCNL1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NALCN | |
Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN | |
0.1 mL | |
259232 | |
Human, Mouse, Rat | |
Purified |
Immunocytochemistry, Immunofluorescence | |
S187-7 | |
Immunocytochemistry/Immunofluorescence | |
bA430M15.1, CanIonFLJ44659, FLJ23913, FLJ44764, four repeat voltage-gated ion channel, MGC74524, sodium leak channel non-selective protein, sodium leak channel, non-selective, VGCNL1, voltage gated channel like 1, Voltage gated channel-like protein 1 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 200kDa. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction