Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584900 Supplier Invitrogen™ Supplier No. PA584900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QALDRPASPP APSGSQQGPE EEAAEALLTE TVRSQTHSLP Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 85%.

VGF was initially identified as a rapidly regulated gene product in nerve growth factor-treated PC12 cells. This protein is synthesized in neurons in the central and peripheral nervous system as well as in the pituitary, adrenal medulla, endocrine cells of the stomach, and pancreatic beta cells. VGF is thought to be involved in organism energy balance and regulation of homeostasis as mice lacking this gene show deficits in these areas. More recent results suggest that VGF is upregulated by brain-derived neurotrophic factor (BDNF) and can stimulate the proliferation of hippocampal progenitor cells and produce antidepressant-like behavioral effects, suggesting that this pathway may be a suitable target for therapeutic treatments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VGF
Gene Accession No. O15240
Gene Alias Antimicrobial peptide VGF[554-577]; Gm1052; NERP-1; NERP-2; Neuroendocrine regulatory peptide-1; Neuroendocrine regulatory peptide-2; neuro-endocrine specific protein VGF; Neurosecretory protein VGF; SCG7; SgVII; Vgf; VGF nerve growth factor inducible; VGF(180-194); VGF(24-63); VGF(375-407); VGF8a protein; VGF-derived peptide AQEE-30; VGF-derived peptide HFHH-10; VGF-derived peptide LQEQ-19; VGF-derived peptide TLQP-11; VGF-derived peptide TLQP-21; VGF-derived peptide TLQP-30; VGF-derived peptide TLQP-62
Gene Symbols VGF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VGF. Recombinant protein control fragment (Product #RP-107883).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7425
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.