Learn More
Invitrogen™ VGLL2 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | VGLL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | VGLL2 |
| Gene Accession No. | Q8BGW8, Q8N8G2 |
| Gene Alias | C130057C21Rik; Protein VITO1; RP1-179P9.2; Transcription cofactor vestigial-like protein 2; Vestigial and Tondu related protein 1; vestigial like 2; vestigial like 2 homolog; vestigial like 2 homolog (Drosophila); vestigial like family member 2; vestigial-like 2; vestigial-like family member 2; Vgl2; vgl-2; Vgll2; Vito1; VITO-1 |
| Gene Symbols | VGLL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.