Learn More
Invitrogen™ VGLL3 Polyclonal Antibody

Description
VGLL3 Polyclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | VGLL3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | VGLL3 |
| Gene Accession No. | A8MV65 |
| Gene Alias | 1700110N18Rik; 4832416J22; C80713; colon carcinoma related protein; LOW QUALITY PROTEIN: transcription cofactor vestigial-like protein 3; RGD1560030; Transcription cofactor vestigial-like protein 3; vestigial like 3; vestigial like 3 (Drosophila); vestigial like family member 3; vestigial-like family member 3; VGL3; VGL-3; Vgll3; Vito-2; zmp:0000000597 |
| Gene Symbols | VGLL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.