Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGLL3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988884
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8884 Supplier Bioss Supplier No. BS9184R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May act as a specific coactivator for the mammalian TEFs.

Specifications

Antigen VGLL3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VGLL3
Gene Accession No. A8MV65
Gene Alias 1700110N18Rik; 4832416J22; C80713; colon carcinoma related protein; LOW QUALITY PROTEIN: transcription cofactor vestigial-like protein 3; RGD1560030; Transcription cofactor vestigial-like protein 3; vestigial like 3; vestigial like 3 (Drosophila); vestigial like family member 3; vestigial-like 3; vestigial-like family member 3; VGL3; Vgl-3; Vgll3; Vito-2
Gene Symbols VGLL3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VGLL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389136
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.