Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558303
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558303 Supplier Invitrogen™ Supplier No. PA558303
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PISPSKRKFS MEPGDEDLDC DNDHVSKMSR IFNPHLNKTA NGDCRRDPRE RSRSPIERAV APTMSLHGSH LYTSLP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

Transcription cofactor vestigial-like protein 4 (VGLL4) acts as a specific coactivator for the mammalian TEFs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLL4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Vgll4
Gene Accession No. Q14135
Gene Alias BC048841; KIAA0121; Transcription cofactor vestigial-like protein 4; vestigial like 4; vestigial like 4 (Drosophila); vestigial like family member 4; Vestigial-like 4; vestigial-like family member 4; VGL-4; Vgll4
Gene Symbols Vgll4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VGLL4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.