Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGLUT1 Antibody, Novus Biologicals™
SDP

Sheep Polyclonal Antibody

$206.50 - $478.00

Specifications

Antigen VGLUT1
Concentration LYOPH
Dilution Western Blot 1:300-1:2000, Flow Cytometry 1: 300 - 1: 2000 Immunohistochemistry 1:300-1:2000
Applications Western Blot, Immunohistochemistry, Flow Cytometry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018408
SDP
View Documents
Novus Biologicals
NBP1202670.025ML
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP120267
SDP
View Documents
Novus Biologicals
NBP120267
0.1 mL
Each for $478.00
Only null left
Add to Cart
 
Description

Description

VGLUT1 Polyclonal specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

VGLUT1
Western Blot 1:300-1:2000, Flow Cytometry 1: 300 - 1: 2000 Immunohistochemistry 1:300-1:2000
Polyclonal
Antisera
RUO
Q62634
57030
IgG
Unpurified
VGLUT1 (both isoforms 1 and 2).
LYOPH
Western Blot, Immunohistochemistry, Flow Cytometry
Unconjugated
Sheep
Human, Mouse, Rat
BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1
SLC17A7
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.