Learn More
VGLUT1 Antibody (S28-9), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259329G
Catalog No. NBP259329G
Only null left
Add to Cart
Description
Description
VGLUT1 Monoclonal specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
VGLUT1
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
SLC17A7
Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1
100 μL
57030
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry
S28-9
Western Blot, Immunohistochemistry
BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1
Mouse
Protein G purified
Primary
Detects 52kDa. No cross-reactivity against VGlut2.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.