Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLUT1 Monoclonal Antibody (A7A3)
GREENER_CHOICE

Catalog No. FEMA550567
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550567 Supplier Invitrogen™ Supplier No. MA550567
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Rat brain tissue lysates, mouse brain tissue lysates. Predicted band size: 62 kDa Subcellular Location: Synaptic vesicle membrane, Membrane, Synaptosome.

A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLUT1
Applications Western Blot
Classification Monoclonal
Clone A7A3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC17A7
Gene Accession No. Q3TXX4, Q62634, Q9P2U7
Gene Alias 2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1
Gene Symbols SLC17A7
Host Species Mouse
Immunogen Synthetic peptide within Human VGluT1 aa 501-550/560.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116638, 57030, 72961
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.