Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLUT1 Monoclonal Antibody (N28/9), FITC
GREENER_CHOICE

Catalog No. PIMA545504
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545504 Supplier Invitrogen™ Supplier No. MA545504
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45504 was sufficient for detection of VGLut1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 52kDa. No cross-reactivity against VGlut2. This antibody was formerly sold as clone S28-9.

A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLUT1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N28/9
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene SLC17A7
Gene Accession No. Q3TXX4, Q62634, Q9P2U7
Gene Alias 2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1
Gene Symbols SLC17A7
Host Species Mouse
Immunogen Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116638, 57030, 72961
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.