Learn More
Invitrogen™ VGLUT1 Monoclonal Antibody (N28/9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527614
Description
1 μg/mL of MA5-27614 was sufficient for detection of VGLut1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 52kDa. No cross-reactivity against VGlut2. This antibody was formerly sold as clone S28-9.
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
Specifications
VGLUT1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Q3TXX4, Q62634, Q9P2U7 | |
SLC17A7 | |
Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (PFA fixed), Western Blot | |
N28/9 | |
Unconjugated | |
SLC17A7 | |
2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1 | |
Mouse | |
Protein G | |
RUO | |
116638, 57030, 72961 | |
-20°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.