Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLUT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588830 Supplier Invitrogen™ Supplier No. PA588830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHKTREEWQY VFLIASLVHY GGVIFYGVFA SGEKQPWAEP EEMSEEKCGF VGHDQLAGSD DSEMEDEAEP PGAPPAPPPS YGATHSTFQP PRPPPPVRDY; Positive Samples: SH-SY5Y; Cellular Location: Cell junction, Cytoplasmic vesicle, Membrane, Multi-pass membrane protein, secretory vesicle, synapse, synaptic vesicle membrane, synaptosome.

A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLUT1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC17A7
Gene Accession No. Q3TXX4, Q9P2U7
Gene Alias 2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1
Gene Symbols SLC17A7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 461-560 of human SLC17A7 (NP_0647051).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57030, 72961
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.