Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGluT1 Rabbit anti-Human, Mouse, Rat, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN164725UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1647-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1647-25UL Supplier MilliporeSigma Supplier No. ABN164725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-VGLUT1, Cat. No. ABN1647, is a highly specific rabbit polyclonal antibody that targets Vesicular glutamate transporter 1 and has been tested in Immunoautoradiography, Immunofluorescence, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGluT1
Applications Immunoradiometric Assay, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide and 50% glycerol.
Gene Alias Vesicular glutamate transporter 1;Brain-specific Na(+)-dependent inorganic phosphate cotransporter;Solute carrier family 17 member 7
Gene Symbols Slc17a7;Bnpi;Vglut1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of rat Vesicular glutamate transporter 1 (VGLUT1).
Molecular Weight of Antigen ∽55 kDa observed; 61.67 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Vesicular glutamate transporter 1 in Human, Mouse, and Rat brain. It targets an epitope with in 14 amino acids from the C-terminal region.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.