Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGLUT2 Antibody (S29-29), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | VGLUT2 |
---|---|
Clone | S29-29 |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
VGLUT2 Monoclonal specifically detects VGLUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
VGLUT2 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
57084 | |
Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2 | |
Primary | |
Detects 60kDa. |
S29-29 | |
Monoclonal | |
Purified | |
RUO | |
differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, differentiation-associated Na-dependent inorganic phosphate cotransporter, DNPI, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2, VGLUT2 | |
SLC17A6 | |
IgG1 | |
Protein G purified | |
60 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title