Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ VGluT2, Guinea Pig, Unlabeled, Polyclonal
GSA_VA

Catalog No. AB2251IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB2251IMI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2251IMI Supplier MilliporeSigma™ Supplier No. AB2251I
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

AB2251-I is a Guinea pig polyclonal antibody that detects VGluT2 and is tested for use in Immunohistochemistry (Paraffin), and Western Blotting.  Vesicular glutamate transporter 2 (UniProt Q9JI12; also known as Differentiation-associated BNPI, Differentiation-associated Na(+)-dependent inorganic phosphate cotransporte, Solute carrier family 17 member 6, VGluT2) is encoded by the Slc17a6 (also known as Dnpi, Vglut2) gene (Gene ID 84487) in rat species. Vesicular glutamate transporters (VGluTs) are proton-dependent carriers responsible for the upload of the major excitatory neurotransmitter glutamate into synaptic vesicles within presynaptic terminals to allow later regulated release at the synaptic cleft. Studies conducted in mice show that VGluT1 and VGluT2 are essential for vital functions, while Vglut3-knockout mice have a normal life expectancy despite being deaf and developing anxiety and discrete basal locomotor phenotypes. VgluT1- and VGluT2-expressing neurons represent the bulk of canonical glutamatergic neurons in the brain, with VgluT1 representing the major subtype and accounting for approximately 80% of total vesicular transport of glutamate in the brain. In contrast, VgluT3 is found in discrete population of non-glutamatergic neurons using other transmitters than glutamate, such as cholinergic interneurons in the dorsal and ventral striatum, subpopulations of GABAergic basket cells in the cortex and hippocampus, and serotoninergic neurons, where it is involved in more subtle modulation of local transmission. Rat VGluT2 is a 12-transmembrane (a.a. 72-92, 126-146, 149-169, 178-198, 217-237, 245-265, 311-331, 350-370, 387-407, 410-430, 444-464, 478-498) protein with 7 cytoplasmic and 6 vacuolar domains, having both its N- and C-terminal tails (a.a. 1-71 and 499-582) exposed at the cytoplasmic side.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen VGluT2
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Guinea pig polyclonal antibody serum with 0.05% Sodium Azide.
Gene Slc17a6, Dnpi, Vglut2
Gene Accession No. Q9JI12
Host Species Guinea Pig
Immunogen KLH-conjugated linear peptide corresponding to the C-terminal sequence of rat VGluT2.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_445879
Test Specificity This polyclonal antibody targets VGluT2 C-terminal end sequence.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.