Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLUT2 Monoclonal Antibody (CL2921)
GREENER_CHOICE

Catalog No. PIMA531378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531378 Supplier Invitrogen™ Supplier No. MA531378
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: CGFIHEDELD EETGDITQNY INYGTTKSYG ATTQANGGWP SGWEKKEEFV QGEVQDSHSY KDRVDY Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 85%.

VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLUT2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL2921
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC17A6
Gene Accession No. Q8BLE7, Q9JI12, Q9P2U8
Gene Alias 2900073D12Rik; differentation-associated Na-dependent inorganic phosphate cotransporter; differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; Dnpi; SLC17A6; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGLUT2
Gene Symbols SLC17A6
Host Species Mouse
Immunogen Recombinant protein fragment of Human SLC17A6 (Product #RP-97279)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140919, 57084, 84487
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.