Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLUT2 Monoclonal Antibody (N29/29), PE
GREENER_CHOICE

Catalog No. PIMA545510
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545510 Supplier Invitrogen™ Supplier No. MA545510
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45510 was sufficient for detection of VGLut2 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 60kDa. This antibody was formerly sold as clone S29-29.

VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLUT2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N29/29
Concentration 1 mg/mL
Conjugate PE
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene SLC17A6
Gene Accession No. Q8BLE7, Q9JI12, Q9P2U8
Gene Alias 2900073D12Rik; differentation-associated Na-dependent inorganic phosphate cotransporter; differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; Dnpi; SLC17A6; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGLUT2
Gene Symbols SLC17A6
Host Species Mouse
Immunogen Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140919, 57084, 84487
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.