Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VGLUT2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143069
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.
Specifications
| VGLUT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC17A6 | |
| 2900073D12Rik; differentation-associated Na-dependent inorganic phosphate cotransporter; differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; Dnpi; SLC17A6; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGLUT2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 57084 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9P2U8 | |
| SLC17A6 | |
| Peptide with sequence C-EDELDEETGDITQN. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction