Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLUT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110380 Supplier Invitrogen™ Supplier No. PA5110380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEKQPWADPE ETSEEKCGFI HEDELDEETG DITQNYINYG TTKSYGATTQ ANGGWPSGWE KKEEFVQGEV QDSHSYKDRV DYS.

VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLUT2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC17A6
Gene Accession No. Q8BLE7, Q9JI12, Q9P2U8
Gene Alias 2900073D12Rik; differentation-associated Na-dependent inorganic phosphate cotransporter; differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; Dnpi; SLC17A6; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGLUT2
Gene Symbols SLC17A6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500-582 of human SLC17A6 (NP_0650791).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140919, 57084, 84487
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.