Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGlut2 Transporter Monoclonal Antibody (S29-29), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 56515100UG

This item is not returnable.
View return policy
Description
VGlut2 Transporter Monoclonal antibody specifically detects VGlut2 Transporter in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western BlotSpecifications
VGlut2 Transporter | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Q8BLE7, Q9JI12, Q9P2U8 | |
Mouse | |
Protein A | |
RUO | |
140919, 57084, 84487 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
S29-29 | |
Unconjugated | |
SLC17A6 | |
2900073D12Rik; differentation-associated Na-dependent inorganic phosphate cotransporter; differentiation-associated BNPI; Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter; differentiation-associated Na-dependent inorganic phosphate cotransporter; Dnpi; SLC17A6; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6; solute carrier family 17 (vesicular glutamate transporter), member 6; solute carrier family 17 member 6; Vesicular glutamate transporter 2; VGlU2; VGLUT2 | |
Fusion protein corresponding to aa 501-582 (cytoplasmic C-terminus) of rat VGlut2 (accession no. NP_445879). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction