Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGluT3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$160.07 - $480.74
Specifications
| Antigen | VGluT3 |
|---|---|
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
VGluT3 Polyclonal antibody specifically detects VGluT3 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
| VGluT3 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8 | |
| SLC17A8 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| 246213 | |
| IgG | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title