Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGluT3 Antibody (S34-34), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259331AF700
Description
VGluT3 Monoclonal specifically detects VGluT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
VGluT3 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SLC17A8 | |
Fusion protein amino acids 546-588 (cytoplasmic C-terminus) of rat VGlut3 | |
100 μL | |
246213 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S34-34 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8 | |
Mouse | |
Protein G purified | |
Primary | |
Detects 65kDa. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction