Learn More
Invitrogen™ VGLUT3 Monoclonal Antibody (N34/34), APC

Description
1 μg/mL of MA5-45514 was sufficient for detection of VGLUT3 in 20 μg of CV-1 fibroblast cells (lysate) transfected with VGLUT3 by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 65kDa. This antibody was formerly sold as clone S34-34.
Specifications
Specifications
| Antigen | VGLUT3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N34/34 |
| Concentration | 1 mg/mL |
| Conjugate | APC |
| Formulation | 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 |
| Gene | SLC17A8 |
| Gene Accession No. | Q7TSF2, Q8BFU8, Q8NDX2 |
| Gene Alias | BC042593; DFNA25; SLC17A8; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8; solute carrier family 17 (vesicular glutamate transporter), member 8; solute carrier family 17 member 8; Vesicular glutamate transporter 3; vesicular glutamate transporter-3; VGLUT 3; VGLUT3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.