Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLUT3 Monoclonal Antibody (N34/34)
GREENER_CHOICE

Catalog No. PIMA527594
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527594 Supplier Invitrogen™ Supplier No. MA527594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

1 μg/mL of MA5-27594 was sufficient for detection of VGLUT3 in 20 μg of CV-1 fibroblast cells (lysate) transfected with VGLUT3 by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 65kDa. This antibody was formerly sold as clone S34-34.

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT-3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLUT3
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N34/34
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene SLC17A8
Gene Accession No. Q7TSF2, Q8BFU8, Q8NDX2
Gene Alias BC042593; DFNA25; SLC17A8; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8; solute carrier family 17 (vesicular glutamate transporter), member 8; solute carrier family 17 member 8; Vesicular glutamate transporter 3; vesicular glutamate transporter-3; VGLUT 3; VGLUT3
Gene Symbols SLC17A8
Host Species Mouse
Immunogen Fusion protein amino acids 546-588 (cytoplasmic C-terminus) of rat VGLUT3.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216227, 246213, 266767
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.