Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VH 032 amide-alkylC4-acid
GREENER_CHOICESDP

Catalog No. 668025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-802-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 66-802-5 Supplier Tocris Bioscience™ Supplier No. 6680/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

VHL ligand with alkyl linker and terminal acid for onward chemistry

VH 032 amide-alkylC4-acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus alkylC4 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

This product has been recently renamed. The previous name for this product was VH 032 - linker 5

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material 6-((-1-((2 S,4 R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-6-oxohexanoic acid
CAS 2172819-74-6
Synonym VH 032 - linker 5
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C28H38N4O6S
Formula Weight 558.69
Product Type Functionalized von-Hippel-Lindau (VHL) Ligand
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.