Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
VH 298 is a high-affinity inhibitor of E3 ubiquitin ligase VHL (Kd = 80-90 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. Results in time- and concentration-dependent accumulation of hydroxylated HIF-α, and upregulates mRNA and protein levels of HIF target genes, with similar transcriptional profile to hypoxia. Cell permeable.
Negative control cis VH 298 also available.
Specifications
Specifications
| Quantity | 10 mg |
| Target | Ubiquitin Ligase (E3) |
| Molecular Formula | C27H33N5O4S |
| Formula Weight | 523.65g/mol |
| Product Type | Inhibitor |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

