Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vibrio Cholera Toxin Alpha Subunit Mouse, Clone: 2H9, Abnova™

Catalog No. 89935082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-082 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-082 Supplier Abnova Corporation Supplier No. MAB12439

Mouse monoclonal antibody raised against Vibrio Cholera Toxin Alpha Subnuit.

Specifications

Antigen Vibrio Cholera Toxin Alpha Subunit
Applications ELISA
Classification Monoclonal
Clone 2H9
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Vibrio Cholera Toxin Alpha Subnuit.
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.