Description
Assure contamination control and increase worker comfort with reusable apparel made from this state-of-the-art, high-performance ESD cleanroom fabric. Designed for the microelectronics industry and any applications where products must be protected from static dissipative charges, airborne particles, and lint. With a super-dense weave of 100% microdenier polyester fibers (over 19,000 filaments per square inch) Burlington™ C-3 meets the highest standard of particulate control. A microengineered carbon fiber grid dissipates electrostatic charges and minimizes manufacturing defects. Made in USA.
- Made of B-Fore fabric, a super-dense weave of 100% microdenier polyester fibers that minimize particulate contamination and a microengineered carbon fiber grid that dissipates electrostatic charges
- Feature a convenient color-coded hanger loop for quick size identification
- With military collar and long raglan sleeves with ESD knit cuffs
- Full-length, limited-linting front zipper with two-ply fly stiffener closure, and snap adjustments at the collar and ankles
- Snaps on back of each leg attach to boots and prevent pullout
Order Info
Available in white,light blue and navy. Made in USA.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 4X-Large |
| Material | Polyester |
| Color | White |
| Disposable | Reusable |
| Closure Type | Zipper |