Learn More
Vidaro™ B-Fore™ ESD Hoods
Catalog No. 19999874 Shop All Vidaro Corporation Products
Change view
Click to view available options
Color:
Light Blue
Navy
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-999-874 Navy Small
19-999-855 Light Blue X-Small
19-999-856 Light Blue Small
19-999-858 Light Blue Large
19-999-860 Light Blue 2X-Large
19-999-873 Navy X-Small
19-999-875 Navy Medium
19-999-876 Navy Large
19-999-877 Navy X-Large
19-999-878 Navy 2X-Large
19-999-883 White X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-999-874 Supplier Vidaro™ Supplier No. H4358/S
This item is not returnable. View return policy
Requires
Two female snaps on each cheek allow the attachment of a facial (not included).
High-performance fabric ideal for critical environments.
  • Super-dense weave of 100% microdenier polyester fibers minimizes particulate contamination
  • Microengineered carbon fiber grid dissipates electrostatic charges
  • Open-faced hood fits snugly under the chin, leaving the entire face exposed
  • Full-bib apron is long, to fit securely into a coverall or frock without pulling out.
  • Front closure has two stainless-steel snaps, and there are adjustment snaps at the back of the neck
Specifications
Product Type Open Face
Size Small
Material Polyester, Carbon
Color Navy
Closure Type Snap Front
Disposable Reusable
Model H-4358
Quantity 1 Ea.
Includes Two snaps on each cheek, Stainless Steel front-snap closure, snap at back of neck for adjustment