Learn More
Vidaro™ B-Fore Pullover ESD Hoods10
Catalog No. 19051378G Shop All Vidaro Corporation Products
Change view
Click to view available options
Color:
Orange
Size:
X-Small
Small
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-051-378G Orange 3X-Large
19-051-378A Orange X-Small
19-051-378B Orange Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-051-378G Supplier Vidaro™ Supplier No. HB395885/3XL (S/306)
This item is not returnable. View return policy
Specifications
Size 3X-Large
Material Polyester, Carbon
Color Orange
Disposable Single-use
Model H-3958-85
Quantity 1 Ea.
Includes Extra long shoulder drape, elastic take up in back with adjustable snap, sewn-in replaceable over the nose 2-ply Poly Tricot facial